ContactUsLinkComponent
3M™ F9469PC VHB™ Adhesive Transfer Tape, Tapecase
3M™ F9469PC VHB™ Adhesive Transfer Tape, Tapecase
  76243-410
3M™ F9469PC VHB™ Adhesive Transfer Tape, Tapecase
  76243-410
 :  F9469PC-12
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    3M F9469PC VHB™ Adhesive Transfer Tape
  • Dimensions:
    12×2160"
  • Cat. No.:
    76243-410
3M™ Adhesive Transfer Tapes are rolls of pressure sensitive adhesive pre-applied to a special release liner. For application, the tape is simply pressed, adhesive side down, to a surface and the liner is peeled off.
  • High holding power in static and dynamic loads
  • Suitable for industrial joining and metal fabrication
  • Eliminates mechanical fastening in many jobs
  • High performance acrylic adhesive