Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Clamps
General Purpose Clamps
Spatter Resist C-Clamp, Zoro
Spatter Resist C-Clamp, Zoro
Spatter Resist C-Clamp, Zoro
Catalog # 76249-568
Supplier:
ZORO - CUSTOMER SERVICE TE
Expand image
Spatter Resist C-Clamp, Zoro
Catalog # 76249-568
Supplier:
ZORO - CUSTOMER SERVICE TE
Supplier Number:
408-P
Specifications
Color:
Gray
Description:
Spatter Resist C-Clamp
Material:
Forged steel
Cat. No.:
76249-568
Finish:
Powder coated
Throat depth:
5"
Specifications
About this item
Product Details
Product Details