Order Entry
United States
ContactUsLinkComponent
Clamps, Holland Applied Technologies
Clamps, Holland Applied Technologies
  76249-326
Clamps, Holland Applied Technologies
  76249-326
 :  13MHHM-0.75

 

  • Description:
    Clamp, Triclover
  • Length:
    0.75"
  • Material:
    Stainless steel 316L
  • Cat. No.:
    76249-326