Aqua-Pure® V-Band Clamp Assembly, 3M
Catalog # 76240-452
Supplier: 3M Healthcare
Specifications
- Description:V-Band Clamp Assembly
- Cat. No.:76240-452
Specifications
About this item
V-band clamp is used for quick and easy opening and closing.
V-band clamp is used for quick and easy opening and closing.