3M™ Circuit Plating Tape, 3M
Supplier: 3M
Jump to:
About this item
3M™ Circuit Plating Tape 1280 with polyester film used for PCB masking tape or other high temperature applications.
- Polyethylene tape core instead of cardboard
- Polyester backing reduces stretch and recovery of tape and associated uneven stop-off line
- Solvent clean up of the board is recommended to facilitate subsequent soldering
- Low stretch polyester film for machine, as well as hand application
- Thick adhesive coating reduces undercutting because adhesive helps block out solutions between fingers
3 Options Available Below
Product Details & Documents
3M™ Circuit Plating Tape 1280 with polyester film used for PCB masking tape or other high temperature applications.
- Polyethylene tape core instead of cardboard
- Polyester backing reduces stretch and recovery of tape and associated uneven stop-off line
- Solvent clean up of the board is recommended to facilitate subsequent soldering
- Low stretch polyester film for machine, as well as hand application
- Thick adhesive coating reduces undercutting because adhesive helps block out solutions between fingers
Product Family Options
Product Information
- ColorAdhesive materialLengthWidthThicknessMaterialTemperature rangeTensile strengthAvailabilityPrice
- 76245-242Polyester tapePolyester tapeRedSilicone/rubber blend2592"2"3.6 mil.Polyester–40...+155 °C30 lbs./in.Each$72.40Case of 24$1,737.60Supplier Number: 7000048445
- 76240-706Polyester tapePolyester tapeRedSilicone/rubber blend2592"1"3.6 mil.Polyester–40...+155 °C30 lbs./in.Each$42.40Case of 36$1,526.40Supplier Number: 7000057465
- 76246-328Polyester tapePolyester tapeRedSilicone/rubber blend2592"½"3.6 mil.Polyester–40...+155 °C30 lbs./in.Each$21.20Case of 72$1,526.40Supplier Number: 7010045078