ContactUsLinkComponent
Cal Adjustable Torque Screwdriver, Sid Tool
Cal Adjustable Torque Screwdriver, Sid Tool
  76245-246
Cal Adjustable Torque Screwdriver, Sid Tool
  76245-246
 :  78329646
  • Description:
    Cal Series Adjustable Torque Screwdriver, CAL 36/4
  • Length:
    7.8"
  • Cat. No.:
    76245-246
  • Weight:
    0.5 lbs.
  • Drive size:
    1/4" F Hex
The work is sophisticated or relatively simple, experienced professionals torque screwdrivers for the industry best accuracy, reliability, and durability. It is ideal for high repeatability for low torque is needed and excellent durability and versatility are required.
  • Short dwell increases through put and reduces repetitive motion injuries
  • Anti-backlash design enhances repeatability
  • Magnifying lens improves the view of clearly engraved scale
  • Clockwise and counter-clockwise operation for true versatility
  • Incredibly durable