Conductive Box with Foam, Menda
Catalog # 76248-506
Supplier: Protektive Pak
Specifications
- Description:Conductive Box with Foam, Molded
- Dimensions:1.3×1.3×0.46" (33×33×12 mm)
- Cat. No.:76248-506
Specifications
About this item
This provides ESD protective storage and handling of ESD sensitive items.
- RTT <1×10 E5 ohms - electrostatic charges able to be removed when grounded
- Secure snap closure keeps boxes from accidentally opening
- Conductive foam inserts provide protection from vibration and shock
ESD protective symbol molded into the cover of the box as per per ESD S541.