Ball Mill, All-Spec Industries
Catalog # 76247-048
Supplier: HISCO, INC TE
Specifications
- Ø:0.019"
- Description:Ball Mill, Carbide
- Size:#1/4
- Cat. No.:76247-048
Specifications
About this item
Carbide tipped ball mills make reaming damaged plated through holes and grinding away damaged baseboard material a snap. The 3/32" shaft fits the micro drill system hand piece and the mill handle.