Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Transfer Tubing
Anti-Static Polyethylene Tubing, Associated Bag
Anti-Static Polyethylene Tubing, Associated Bag
Anti-Static Polyethylene Tubing, Associated Bag
Catalog # 76250-786
Supplier:
Associated Bag
Expand image
Anti-Static Polyethylene Tubing, Associated Bag
Catalog # 76250-786
Supplier:
Associated Bag
Supplier Number:
SP-427-0161462
Specifications
Tubing ext.Ø:
14"
Description:
Anti-static Tubing
Length:
22"
Cat. No.:
76250-786
Specifications
About this item
Product Details
Product Details