ContactUsLinkComponent
3M™ Aluminum Foil Shielding Tape 1170, Budnick Converting
3M™ Aluminum Foil Shielding Tape 1170, Budnick Converting
Catalog # 76268-050
Supplier:  Budnick Converting
3M™ Aluminum Foil Shielding Tape 1170, Budnick Converting
Catalog # 76268-050
Supplier:  Budnick Converting
Supplier Number:  1170M2400W2900L
Specifications
  • Dimensions:
    2.4×2.9"
  • Cat. No.:
    76268-050
Specifications
About this item
Aluminum foil shielding tape 1170 with with a electrically conductive, acrylic adhesive that ensures firm adherence. Flame-retardant tape is designed for applications that require reliable, point-to-point electrical contact.
  • Pressure-sensitive tapes, that the surface to which the tape is applied be clean, dry and free of grease and oil
Shelf life from date of manufacture is based on the product being stored at 70 °F(21 °C)/50% relative humidity out of direct sunlight.