Order Entry
United States
ContactUsLinkComponent
Pneumatic Applicator with Dual Cartridge, Zoro
Pneumatic Applicator with Dual Cartridge, Zoro
Catalog # 76253-286
Pneumatic Applicator with Dual Cartridge, Zoro
Catalog # 76253-286
Supplier Number:  G6123625

Specifications

  • Color:
    Black
  • Description:
    Pneumatic Adhesive Dispenser,50 ml Dual Cartridges
  • Cat. No.:
    76253-286
  • For use with:
    Two-Component Epoxies, Urethanes, Methacrylates

Specifications

About this item

This is for pneumatic adhesive dispensing, 1:1 and 2:1 mix ratio products in 50 ml Dual cartridge.