Standard Hand Tap, Sid Tool
Catalog # 76249-004
Supplier: SID TOOL CO INC TE
Specifications
- Description:Straight Flute Standard Hand Tap, with M18×1.00 thread
- Length:4¹⁄₃₂"
- Cat. No.:76249-004
- Thread length:1¹³⁄₁₆"
Specifications
About this item
Straight four flute standard hand tap, with bottoming chamfers are used for threading blind holes. They have 1 to 2 threads.
- High speed steel (HSS) provides good wear-resistance
- Used in general purpose applications for both ferrous and nonferrous materials
- Bright provides a smooth, polished finish on the tool
It increases chip flow in softer materials such as aluminum, wood and plastic.