Adapter Assemblies, Apex Tools
Supplier: Cooper Tools
Jump to:
About this item
Syringe adapter assembly have a unique design works with syringes.
- Plugs directly into any Kahnetics shot meter
- Swivel 360° on adapter head for ease of motion
Produces repeatable shots of material and requires minimal clean-up.
1 Options Available Below
Product Details & Documents
Syringe adapter assembly have a unique design works with syringes.
- Plugs directly into any Kahnetics shot meter
- Swivel 360° on adapter head for ease of motion
Produces repeatable shots of material and requires minimal clean-up.
Product Family Options
Product Information
- AvailabilityPrice
- 76253-450Syringe Adapter Assembly, Plastic, 5 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated SyringesSyringe Adapter Assembly, Plastic, 5 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated SyringesLog in for availabilityEach$166.12Supplier Number: KDS505S6N
- 76253-452Syringe Adapter Assembly, Plastic, 30 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated SyringesSyringe Adapter Assembly, Plastic, 30 cc, 3/32" tube I.D., for use with Luer Lok™, Luer Slip™ and Tapered Tip Air Operated SyringesDiscontinued-Supplier Number: -