3M™ VHB™ Tapes, 3M
Supplier: 3M
Certificates
About this item
These tapes utilizes multi-purpose acrylic adhesive on both sides of a conformable, foam core. Also replaces mechanical fasteners (rivets, welds, screws) or liquid adhesives.
- Fast and easy-to-use permanent bonding method provides high strength and long-term durability
- Virtually invisible fastening keeps surfaces smooth
- Eliminate drilling, grinding, refinishing, screwing, welding and associated clean-up
- Creates a permanent seal against water, moisture and more
- Pressure sensitive adhesive bonds on contact to provide immediate handling strength
- Allows the use of thinner, lighter weight and dissimilar materials
The combination of strength, conformability and adhesion makes this family one of the most capable and well-rounded 3M™ VHB™ Tapes. It is specifically designed for good adhesion to metals, glass and other medium to high surface energy materials. Applications for this tape include stiffener bonding, sealing polycarbonate lens over LCD, windows to painted control panels and mounting vinyl wiring ducts and conduit channels.
Certifications: UL GREENGUARD and UL GREENGUARD Gold Certified, contributing to LEED.
Frequently Bought Together