Extension Support Rings, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Extension support for holding funnels.
- Steel rod, zinc plated
- 12" long
Steel rod, zinc-plated extension support rings. Rod diameter 5/16", overall length 12" for all sizes.
Frequently Bought Together