Order Entry
United States
ContactUsLinkComponent
EnzyChrom™ Gliadin ELISA Assay Kit, BioAssay Systems
EnzyChrom™ Gliadin ELISA Assay Kit, BioAssay Systems
Catalog # 76293-892
Supplier:  Avantor
CAS Number:  
undefined
EnzyChrom™ Gliadin ELISA Assay Kit, BioAssay Systems
Catalog # 76293-892
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Description:
    Gliadin ELISA Assay Kit
  • Size:
    100 assays
  • Cat. No.:
    76293-892

Specifications

About this item

Determination of gliadin in food and beverage samples.

  • Fast and Simple: entire assay procedure takes less than three hours
  • Sensitive and Accurate: quantifies gliadin in the parts per billion range
  • Simple Calculation: simple calculation with linear standard curve
  • Method: Sandwich ELISA (OD450 nm)
  • Samples: Food and beverage samples (e.g. bread, beer, etc)

LIADIN and glutenin are the two main proteins comprising gluten, a protein composite in the endosperm of grains. An intolerance to these gluten proteins results in celiac disease, a chronic autoimmune intestinal disorder characterized by bloating, diarrhea, and other abdominal pains. In addition to celiac disease, there are other forms of gluten intolerance that have led to the adoption of many gluten free alternatives in the food and beverage industry. Different countries have different restrictions regarding what foods can be labeled “gluten free” for gluten sensitive individuals. This has resulted in a need for accurate gluten quantification in food and beverage samples. Fortunately because the gliadin and glutenin fractions of gluten are almost evenly split, quantifying the total content of one of these proteins allows for accurate estimation of total gluten content in a sample.

BioAssay Systems Gliadin ELISA uses a colorimetric sandwich ELISA method detailed in the figure below to accurately measure gliadin content in samples, which can then be used as an accurate estimate of total gluten content.