- Description:Glucose Assay Kit III
- Size:100 assays
- Cat. No.:76293-890
For quantitative determination of glucose and evaluation of drug effects on glucose metabolism.
- Sensitive and accurate: use as little as 20 μl samples; linear detection range in 96-well plate - 0.03 to 2 mM (0.54 mg/dl to 36 mg/dl) glucose
- Simple and high-throughput: the procedure involves addition of a single working reagent and incubation for 30 min at room temperature
- Method: OD565 nm
- Samples: Serum, plasma, urine, saliva, milk, culture medium, food, beverages etc
Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders. Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism. Simple, direct and high-throughput assays for measuring glucose concentrations find wide applications in research and drug discovery.
BioAssay Systems glucose assay kit III uses a single Working Reagent that combines the enzyme reaction and color reaction in one step. The color intensity of the reaction product at 565 nm is directly proportional to the glucose concentration in the sample.