ContactUsLinkComponent
Trio.Bas™ Maxi Holder Cart, Hardy Diagnostics
Trio.Bas™ Maxi Holder Cart, Hardy Diagnostics
Catalog # 76288-234
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
undefined
Trio.Bas™ Maxi Holder Cart, Hardy Diagnostics
Catalog # 76288-234
Supplier:  Hardy Diagnostics
Supplier Number:  BAS372
Product available on GSA Advantage®
Specifications
  • Description:
    Trio.Bas™ Maxi Holder Cart
  • Dimensions:
    25×35×70cm
  • Material:
    Stainless steel
  • Cat. No.:
    76288-234
Specifications
About this item
A multi holder cart with fixed vertical hook holder and extension pole for instruments.
  • Convenient for using one or multiple air samplers and for transportation
Multi Holder Maxi cart is a stainless-steel multi-purpose mobile cart, suitable to position instrument(s) with a vertical hook or on the extension pole. The extension pole can reach to a desired maximum of 6' and minimum of 4'.