Trio.Bas™ CaliTest for Cable Air Sampling Instruments, Hardy Diagnostics
76288-232
Product available on GSA Advantage®
- Description:Trio.Bas™ CaliTest
- Cat. No.:76288-232
A manual verification instrument to check the precision of air flow rates.
- For control only, not designed to take the place of annual calibrations as required by USP
The CaliTest system is for use only with instruments that have direct data transfer cables.