Mouse Recombinant FGFR2/CD332 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse FGFR2 (AAA39377.1) (Met1-Glu263) was expressed with the Fc region of human IgG1 at the C-terminus.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to inhibit FGF-acidic dependent proliferation of Balb/c 3T3 mouse fibroblasts. The ED50 for this effect is typically 0.4-2.4 ng/ml
- Gene ID:AAA39377.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TK14|ECT1|TK25|BEK|JWS|KGFR|CFD1|CEK3|CD332|K-SAM|BBDS|BFR-1
- Protein/Peptide Name:FGFR2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse FGFR2 comprises 483 amino acids and has a predicted molecular mass of 54.1 kDa. The apparent molecular mass of the protein is approximately 89.9 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Glu263
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together