Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant EphB6 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant EphB6 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant EphB6 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Immobilized mouse EPHB6-His at 10 μg/ml (100 μl/well) can bind mouse EFNB1-Fc (Cat:50580-M02H), The EC50 of mouse EFNB1-Fc (Cat:50580-M02H) is 0.12-0.28 μg/ml.
    2. Immobilized mouse EPHB6-His at 10 μg/ml (100 μl/well) can bind mouse EFNB2-Fc (Cat:50598-M02H), The EC50 of mouse EFNB2-Fc (Cat:50598-M02H) is 0.04-0.08 μg/ml.
  • Gene ID:
    O08644-1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Mep|Cekl
  • Protein/Peptide Name:
    EphB6
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse EPHB6 comprises 566 amino acids and has a predicted molecular mass of 61.1 kDa. The apparent molecular mass of the protein is approximately 91 and 65 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ser587
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together