Mouse Recombinant TFRC (from HEK293 cells), His Tag
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the extracellular domain of mouse TFRC (Q62351) (Cys 89-Phe 763) was fused with a polyhistidine tag at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage Conditions:–20…–80 °C
- Endotoxin Content:< 1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:Q62351
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TRFR|T9|TFR1|CD71|p90|TFR|TR
- Protein/Peptide Name:TFRC
- Purity:>92% as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse TFRC comprises 694 amino acids and predicts a molecular mass of 78.3 kDa. The apparent molecular mass of mouse TFRC is approximately 80 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Cys89-Phe763
- Formulation:Lyophilized from sterile 20 mM Tris, 150 mM NaCl, 5% Mannitol, 0.1% tween80, pH 7.5
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together