Mouse Recombinant CD69/CLEC2C/AIM Protein (Fc Tag, ECD) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse Cd69 (NP_001028294.1) (Gly64-Arg199) was expressed with the Fc region of human IgG1 at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_001028294.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
- Protein/Peptide Name:CD69
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse Cd69 consists 396 amino acids and predicts a molecular mass of 44.1 kDa.
- Sequence:Gly64-Arg199
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together