Mouse Recombinant METAP2/MAP2 Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse METAP2 (O08663) (Ala 2-Tyr 478) was expressed,with a C-terminal polyhistidine tag.
- Shipping temperature: Dry ice
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 1180-SE). The specific activity is >15 pmol/min/µg.
- Gene ID:O08663
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:MNPEP|MAP2|p67eIF2|p67
- Protein/Peptide Name:METAP2
- Purity:>88% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse METAP2 consists of 488 amino acids and has a calculated molecular mass of 54.3 kDa. It migrates as an approximately 60 KDa band in SDS-PAGE under reducing conditions.
- Sequence:Ala2-Tyr478
- Shipping Temperature:Ambient
Specifications
Related Information
Frequently Bought Together