Mouse Recombinant CD38 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the extracellular domain of mouse CD38 (NP_031672.2) (Leu 45-Thr 304) was expressed, fused with a C-terminal polyhistidine tag.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to convert the substrate nicotinamide guanine dinucleotide (NGD+) to cyclic GDPribose. The specific activity is >50,000pmols/min/μg.
- Gene ID:NP_031672.2
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ADPRC 1-rs1|Cd38|ADPRC1|Cd38-rs1|I-19
- Protein/Peptide Name:CD38
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse CD38 consists of 271 amino acids and has a calculated molecular mass of 31.3 kDa. As a result of glycosylation, the apparent molecular mass of rmCD38 is approximately 38-42 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Thr304
- Formulation:Lyophilized from sterile 20 mM MES, 0.15 M NaCl, pH 6.5
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together