Mouse Recombinant CD1D/R3G1 Protein (His & Fc Tag) (from HEK293 Cells)
A DNA sequence encoding the extracellular domain of mouse CD1D (NP_031665.2) (Met 1-Gly 305) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_031665.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Ly-38|CD1.1|AI747460|Cd1d|Cd1a
- Protein/Peptide Name:CD1D
- Purity:>80% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse CD1D /Fc is a disulfide-linked homodimeric protein. The reduced monomer consists of 532 amino acids and has a predicted molecular mass of 60.4 kDa.
- Sequence:Met1-Gly305
- Shipping Temperature:Ambient
Frequently Bought Together