Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant PIGR Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant PIGR Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant PIGR Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA
    . Immobilized recombinant mouse PIGR at 5 μg/ml (100 μl/well) can bind mouse IgM with a linear range of 0.156-10 μg/ml
    .
  • Gene ID:
    NP_035212.2
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MGC125362|MGC125361|FLJ22667|PIGR
  • Protein/Peptide Name:
    PIGR
  • Purity:
    >97% as determined by SDS-PAGE
  • Molecular Weight:
    The secreted recombinant mouse PIGR consists of 638 amino acids and has a calculated molecular mass of 70.8 kDa. As a result of glycosylation, the recombinant protein migrates as an approximately 90-100 kDa protein in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Lys645
  • Formulation:
    Lyophilized from sterile PBS, pH 7.4
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together