Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL18/IL-18 Protein (from E. coli)
Mouse Recombinant IL18/IL-18 Protein (from E. coli)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant IL18/IL-18 Protein (from E. coli)
Supplier:  Sino Biological
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to induce IFN-γ secretion by KG-1 human myelomonocyte.
    The ED50 for this effect is typically 0.1-0.8 μg/ml.
  • Gene ID:
    P70380-1
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    IL-1g|IL1F4|IGIF|Interleukin 18|IL-18
  • Protein/Peptide Name:
    IL18
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant mouse IL18 consists of 158 amino acids and predicts a molecular mass of 18.2 KDa. It migrates as an approximately 19 KDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Asn36-Ser192
  • Formulation:
    Lyophilized from sterile PBS, pH 7.4
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together