Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL13RA2/CD213A2 Protein (His & Fc Tag) (from HEK293 Cells)
Mouse Recombinant IL13RA2/CD213A2 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant IL13RA2/CD213A2 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Immobilized human IL13 at 20 μg/ml (100 μl/well) can bind mouse IL13RA2 with a linear range of 0.5-2 μg/ml.
    2. Measured by its ability to inhibit IL13-dependent proliferation of TF1 human erythroleukemic cells. The ED50 for this effect is typically 5-30 ng/ml.
  • Gene ID:
    NP_032382.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD213A2|IL-13R|CT19|IL13BP
  • Protein/Peptide Name:
    IL13Ra2
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse IL13Rα2/Fc is a disulfide-linked homodimer after removal of the signal peptide. The reduced monomer consists of 561 amino acids and has a predicted molecular mass of 64.5 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rmIL13Rα2/Fc monomer is approximately 75-80 kDa due to glycosylation.
  • Sequence:
    Met1-Lys334
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together