Mouse Recombinant CA14 (from HEK293 cells), His Tag
Supplier: Avantor
Certificates
About this item
A DNA sequence encoding the extracellular domain of mouse CA14 (NP_035927.1) (Met 1-Met 290) was expressed with a C-terminal polyhistidine tag.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage Conditions:Store it under sterile conditions at –20 °C to –80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:< 1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its esterase activity. The specific activity is >400 pmoles/min/μg, as measured with 1 mM 4-Nitrophenyl acetate and 0.8 μg enzyme at 400 nm in 100 μL of 12.5 mM Tris, 75 mM NaCl, pH 7.5.
- Gene ID:NP_035927.1
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Protein/Peptide Name:CA14
- Purity:> 95 % as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse CA14 consists of 286 amino acids and has a calculated molecular mass of 32.2 kDa. As a result of glycosylation, the recombinant protein migrates as an approximately 45-48 kDa protein in SDS-PAGE under reducing conditions.
- Sequence:Met1-Met290
- Formulation:Lyophilized from sterile 25mM Tris, 0.15mM NaCl, pH 7.5
- Shipping Temperature:In general, recombinant proteins are provided as lyophilized powder which are shipped at ambient temperature. Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.
Specifications
Frequently Bought Together