AcmNPV Recombinant Envelope glycoprotein gp64/AcmNPV-gp64 Protein (His Tag) (from HEK293 Cells)
Catalog # 103663-650
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:AcMNPV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_054158.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:gp64
- Protein/Peptide Name:AcMNPV-GP64
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant autographa californica nucleopolyhedrovirus (AcNPV)(strain E2) gp64 consists of 472 amino acids and predicts a molecular mass of 54.2 kDa.
- Sequence:Met1-Thr481
- Shipping Temperature:Ambient
- Cat. No.:103663-650
Specifications
About this item
A DNA sequence encoding the autographa californica nucleopolyhedrovirus (AcNPV)(strain E2) gp64 (NP_054158.1) (Met1-Thr481) was expressed with a polyhistidine tag at the C-terminus.