Order Entry
United States
ContactUsLinkComponent
MERS-CoV Recombinant Spike Protein (S1+S2 ECD, aa 1-1297, His Tag) (from Baculovirus Insect Cells)
MERS-CoV Recombinant Spike Protein (S1+S2 ECD, aa 1-1297, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-604
Supplier:  Sino Biological
CAS Number:  
undefined
MERS-CoV Recombinant Spike Protein (S1+S2 ECD, aa 1-1297, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-604
Supplier:  Sino Biological
Supplier Number:  40069-V08B-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    MERS-CoV
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
    1.Immobilized Sh(2C-EMC)(1-1297)(Cat:40069-V08B) at 10 μg/ml (100 μl/well) can bind biotinylated DPP4 (Cat:10688-HNCH),The EC50 of can biotinylated DPP4 (Cat:10688-HNCH) is 0.02-0.04 µg/ml.
    2.Immobilized Sh(2C-EMC)(1-1297)(Cat:40069-V08B) at 10 μg/ml (100 μl/well) can bind biotinylated Fc-DPP4 (Cat:10688-H01H),The EC50 of can biotinylated Fc-DPP4 (Cat:10688-H01H) is 0.01 - 0.02 µg/ml.
  • Gene ID:
    AFS88936.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
  • Protein/Peptide Name:
    Spike
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant extracellular domain of spike protein MERS-CoV comprises 1291 amino acids and has a predicted molecular mass of 142.52 kDa.
  • Sequence:
    Met1-Trp1297
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103663-604

Specifications

About this item

A DNA sequence encoding the MERS-CoV extracellular domain of spike protein (AFS88936.1) (Met1-Trp1297) was fused with a polyhistidine tag at the C-terminus.