Human Recombinant respiratory syncytial virus (RSV) Fusion/RSV-F (Strain RSS-2) Protein (His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-560
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:RSV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P11209
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:fusion protein|HRSVgp08|F
- Protein/Peptide Name:RSV-F
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human RSV (strain RSS-2) consists of 519 amino acids and predicts a molecular mass of 57.8 kDa. The RSV F0 precursor protein is cleaved into the disulfide-linked F1 and F2 subunits. As a result of glycosylation, the apparent molecular mass of the F0 and F1 is approximately 63 kDa and 44 - 53 KDa in SDS-PAGE under reducing conditions, respectively.
- Sequence:Met1-Thr529
- Shipping Temperature:Ambient
- Cat. No.:103663-560
Specifications
About this item
A DNA sequence encoding the extracellular domain of human RSV (strain RSS-2) fusion protein (P11209) (Met 1-Thr 529) was expressed with a polyhistidine tag at the C-terminus.