Human Recombinant Vasoactive intestinal peptide/VIP Protein (His Tag) (from HEK293 Cells)
A DNA sequence encoding the human VIP (P01282-2) (Met1-Lys169) with a C-terminal polyhistidine tag was expressed.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P01282-2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:PHM27
- Protein/Peptide Name:VIP
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human VIP comprises 160 amino acids and has a predicted molecular mass of 18.3 kDa. The apparent molecular mass of the protein is approximately 25, 21 and 15 kDa in SDS-PAGE under reducing conditions
- Sequence:Met1-Lys169
- Shipping Temperature:Ambient
Frequently Bought Together