Human Recombinant RSV (B1) glycoprotein G/RSV-G Protein (His Tag) (from HEK293 Cells)
103623-072
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:RSV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:O36633-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Glycoprotein
- Protein/Peptide Name:RSV-G
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant human RSV strain B1 glycoprotein G comprises 244 amino acids with a predicted molecular mass of 27 kDa. As a result of highly glycosylation, the apparent molecular mass of the recombinant protein is approximately 80-90 kDa in SDS-PAGE under reducing conditions.
- Sequence:His67-Ala299
- Shipping Temperature:Ambient
- Cat. No.:103623-072
A DNA sequence encoding the extracellular domain of human RSV strain B1 glycoprotein G (O36633-1) (His 67-Ala 299) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.