Human Recombinant EZR/VIL2/Ezrin Protein (His Tag) (from E. coli)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mature form of human EZR (P15311) (Pro2-Leu586) was expressed with a polyhistide tag at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information
- Gene ID:P15311
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HEL-S-105|VIL2|CVIL|CVL
- Protein/Peptide Name:EZR
- Purity:>60% as determined by SDS-PAGE
- Molecular Weight:The recombinant human EZR consists of 601 amino acids and predicts a molecular mass of 71.2 KDa. It migrates as an approximately 82 KDa band in SDS-PAGE under reducing conditions.
- Sequence:Pro2-Leu586
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together