Human Recombinant CD47 (from HEK293 Cells)
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with six amino acids (LEVLFQ) at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind human SIRPA-His(Cat:11612-H08H)in functional ELISA. Measured by its ability to bind mouse SIRPA-His(Cat:50956-M08H)in functional ELISA.
- Gene ID:NP_942088.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:IAP|MER6|OA3
- Protein/Peptide Name:CD47
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human CD47 consists 128 amino acids and predicts a molecular mass of 14.5 kDa. The apparent molecular mass of the protein is approximately 35 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Met1-Pro139
- Shipping Temperature:Ambient
- Formulation:Lyophilized from sterile PBS, pH 7.4
Frequently Bought Together