Human Recombinant PTH1R/PTHR1/PTH1 Receptor Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human PTH1R (Q03431) extracellular domain (Met 1-Gly 188) was fused with a polyhistidine tag at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:Q03431
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:PFE|MGC138426|PTHR1|PTHR|MGC138452
- Protein/Peptide Name:PTH1R
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human PTH1R consists of 173 amino acids and has a predicted molecular mass of 20 kDa. As a result of glycosylation, the apparent molecular mass of rh PTH1R is approximately 30-40 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gly188
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together