Human Recombinant CD19 (from HEK293 cells), His Tag
A DNA sequence encoding the human CD19 (P15391) extracellular domain (Met 1-Lys 291) was expressed, with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized Human CD19 His (Cat: 11880-H08H) at 2 μg/ml (100 μl/well) can bind Anti-CD19 Antibody (Cat: 11880-RP01), the EC50 of Anti-CD19 Antibody is 5-40 ng/ml.
- Gene ID:P15391
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CVID3|B4
- Protein/Peptide Name:CD19
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human CD19 consists of 283 amino acids and predictes a molecular mass of 31.6 kDa. In SDS-PAGE under reducing conditions, rhCD19 migrates as an approximately 47 kDa band due to glycosylation.
- Sequence:Met1-Lys291
- Formulation:Lyophilized from sterile PBS, pH 7.4
- Shipping Temperature:Ambient
Frequently Bought Together