Human Recombinant PDE1B Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human PDE1B long isoform (Q01064-1) (Met 1-Asp 536) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:Q01064-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:PDE1B1|PDES1B
- Protein/Peptide Name:PDE1B
- Purity:>94% as determined by SDS-PAGE
- Molecular Weight:The recombinant human PDE1B/GST chimera consists of 773 amino acids and has a calculated molecular mass of 89.2 kDa. It migrates as an approximately 75 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Asp536
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together