Human Recombinant R-Spondin 1/RSPO1 Protein (from CHO cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human RSPO1 (NP_001033722.1) (Met1-Ala263) was expressed and purified.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to induce activation of ßcatenin response in a Topflash Luciferase assay using HEK293T human embryonic kidney cells. The ED50 for this effect is typically 20-120 ng/ml in the presence of 5 ng/ml recombinant mouse Wnt3a.
- Gene ID:NP_001033722.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CRISTIN3|R-Spondin 1|RSPO
- Protein/Peptide Name:RSPO1
- Purity:≥95% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.
- Molecular Weight:The recombinant human RSPO1 consists of 243 amino acids and has a predicted molecular mass of 26.8 kDa. As a result of glycosylation, the apparent molecular mass of it is approximately 40 and 31 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met 1-Ala 263
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together