RSV Recombinant RSV-G (from HEK293 cells), His Tag
103680-502
: Avantor
:
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:RSV
- Size:20 µg
- Tag sequence:His Tag
- Storage Conditions:Store it under sterile conditions at –20...–80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P27022-1
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Protein/Peptide Name:RSV-G
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant human RSV-G comprises 242 amino acids with a predicted molecular mass of 26.7 kDa. As a result of highly glycosylation, the RSV-G protein migrates as an approximately 80-90 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Asn66-Arg297
- Formulation:Lyophilized from sterile PBS, pH 7.4
- Shipping Temperature:In general, recombinant proteins are provided as lyophilized powder which are shipped at ambient temperature. Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.
- Cat. No.:103680-502
A DNA sequence encoding the glycoprotein G extracellular domain (Asn 66-Arg 297) of human respiratory syncytial virus A (95 % homologous with strain rsb1734) (P27022-1) was expressed, with a C-terminal polyhistidine tag.