Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD48/SLAMF2/BCM1 Protein (Fc Tag) (from HEK293 Cells)
Human Recombinant CD48/SLAMF2/BCM1 Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant CD48/SLAMF2/BCM1 Protein (Fc Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its binding ability in a functional ELISA.
    2. Immobilized human 2B4-His (Cat:10042-H08H), at 10 μg/ml (100 μl/well) can bind human CD48-Fc. The EC50 of human CD48-Fc is 0.03-0.06 μg/ml.
  • Gene ID:
    NP_001769.2
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MEM-102|BLAST|hCD48|mCD48|BLAST1|BCM1|Blast-1|SLAMF2
  • Protein/Peptide Name:
    CD48
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human CD48/Fc comprises 432 amino acids and has a predicted molecular mass of 49.1 kDa. The apparent molecular mass of the protein is approximately 64.5 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ser220
  • Formulation:
    Lyophilized from sterile PBS, pH 7.4
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together