Human Recombinant FAP (from HEK293 cells), His Tag
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human FAP isoform 1 (Q12884-1) extracellular domain (Leu 26-Asp 760) was fused with the polyhistidie-tag at the N-terminus.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to convert the substrate benzyloxycarbonyl-Gly-Pro-7-amido-4-methylcoumarin (Z-GP-AMC) to Z-Gly-Pro and 7-amino-4-methylcoumarin (AMC).
The specific activity is >1200 pmol/min/μg. - Gene ID:Q12884-1
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Fapalpha|SIMP|DPPIVA|DPPIV|Fibroblast Activation Protein alpha|FAPA
- Protein/Peptide Name:FAP
- Purity:>95% as determined by SDS-PAGE, ≥90% as determined by SEC-HPLC
- Molecular Weight:The recombinant human FAP consists of 751 amino acids and predicts a molecular mass of 87.2 kDa. As a result of glycosylation, rh FAP migrates as approximately 90 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Leu26-Asp760
- Formulation:Supplied as sterile 25 mM Tris, 250 mM NaCl, pH 8.2
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together