Human Recombinant FCGR2A (from HEK293 cells), His Tag
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human CD32a (AAA35827.1) precursor was expressed, with a C-terminal polyhistidine tag. It is identical to FCGR2A131H/R in the reference.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:1. Measured by its ability to bind biotinylated human IgG2 (Cat: 13504-HNAH) in functional ELISA.
2. Loaded Anti-CD32A Antibody, Mouse Monoclonal(Cat. 10374-MM02) on AMC Biosensor, can bind CD32A Protein, Human, Recombinant (167 Arg, His Tag)(Cat. 10374-H08H) with an affinity constant of 6.8 nM as determined by Octet RED System. - Gene ID:P12318-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD32|FcGR|CD32A|IGFR2|Fc gamma RIIA|FCG2|FCGR2A1|CDw32|FCGR2
- Protein/Peptide Name:FCGR2A
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
- Molecular Weight:The recombinant human FCGR2A consists of 196 amino acids after removal of the signal peptide and predicts a molecular mass of 22 kDa. It migrates as an approximately 30 kDa protein due to glycosylation by SDS-PAGE under reducing conditions.
- Sequence:Met1-Ile218,167R
- Shipping Temperature:Ambient
- Protein Function:Immunotherapy
- Tag sequence:His Tag
- Formulation:Lyophilized from sterile PBS, pH 7.5
Specifications
Frequently Bought Together