Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant Pleiotrophin/PTN/HB-GAM Protein (from Baculovirus Insect cells)
 
undefined
Human Recombinant Pleiotrophin/PTN/HB-GAM Protein (from Baculovirus Insect cells)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized human PTN (Cat:10288-HNAB) at 10 μg/ml (100 μl/well) can bind rat SDC1-Fc (Cat:80344-R02H), The EC50 of rat SDC1-Fc (Cat:80344-R02H) is 0.35-0.81 μg/ml.
  • Gene ID:
    NP_002816.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    HBNF|OSF|HARP|HBBN|Osf1|HB-GAM|Osf-1|HBGF-8
  • Protein/Peptide Name:
    PTN
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human PTN consists of 136 amino acids and predicts a molecular mass of 15.3 KDa. It migrates as an approximately 19 KDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Asp168
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together