Human Recombinant PVR (from HEK293 Cells), His Tag
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human CD155 isoform a (NP_006496.3) extracellular domain (Met 1-Asn 343) was fused with a polyhistidine tag at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized human CD155 (Cat: 10109-H08H) at 2 μg/ml (100 μl/well) can bind human DNAM1 (Cat: 10565-H03H), the EC50 of human DNAM1/CD226 is 50-300 ng/ml.
- Gene ID:P15151-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TAGE4|PVS|CD155|NECL5|Necl-5|HVED
- Protein/Peptide Name:PVR
- Purity:≥97% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.
- Molecular Weight:The recombinant human CD155 consists of 334 amino acids and has a predicted molecular mass of 36.5 kDa. As a result of glycosylation, the rhCD155/Fc monomer migrates as approximately 60-65 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Asn343
- Shipping Temperature:Ambient
- Tag sequence:His Tag
- Formulation:Lyophilized from sterile PBS, pH 7.5
Specifications
Frequently Bought Together