ContactUsLinkComponent
Garage Physics Ballista Kit, Eisco Scientific
Garage Physics Ballista Kit, Eisco Scientific
Catalog # 470307-348
Supplier:  Wards
undefined
Garage Physics Ballista Kit, Eisco Scientific
Catalog # 470307-348
Supplier:  Wards
Supplier Number:  GP00016
Specifications
  • Description:
    Ballista Kit
  • Cat. No.:
    470307-348
Specifications
About this item
Uses torsion spring bundles and levers to launch ping pong balls.
  • No tools needed
  • Pre-cut pieces
  • Allows for multiple launch angles
Ballista Kit is assembled from pre-cut pieces with no tools. Uses torsion spring bundles and levers to launch a ping pong ball up to 30 feet. Allows for multiple launch angles and exploration of projectile motion and trajectories.
Delivery: Includes 12 part kit, string, leather pouch, ping pong ball and guide.