Thermometer Adapter, 10° Offset, Chemglass
80066-102
Small Business Enterprise
- Description:Thermometer Adapter, 10° Offset
- Cat. No.:80066-102
- [ST] inner joint:24/40
- Supplier no.:CG-1054-01
Adapter has a 10/30 outer joint at the top for use with thermometers.
- Allows angling of thermometers into bottom of the flask
- 10° offset
When used with vertical side neck flasks, thermometers can be angled into the bottom of the flask.