Inlet Thermometer Adapters, 10° Offset, Chemglass
Supplier: Chemglass
Certificates
About this item
Adapters have a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7 mm.
- Adapter allows thermometers or bleed tubes to be angled into the bottom of flasks
When used with vertical side neck flasks, thermometers or bleed tubes can be angled into the bottom of the flask.
Delivery: Supplied complete with a compression cap and Viton O-Ring.
Frequently Bought Together